Assay Details
Target Gene Details
Entrez Gene ID: | 54967 |
Gene Name: | cancer/testis antigen 55 |
Gene Aliases: |
BJHCC20A, CXorf48 |
Location: |
Chr.X:135156536-135171827 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CT55 | NM_001031705.2 | NP_001026875.1 | ||
| NM_017863.2 | NP_060333.1 | |||
| XM_005262427.1 | XP_005262484.1 | |||
| AF520814.2 | AAP47205.1 | |||
| AK000534.1 | BAA91237.1 | |||
| AY352211.1 | AAQ90022.1 | |||
| BC020662.1 | AAH20662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469825 | Chr.X:135112233 - 135264658 on Build GRCh38 | Loss |
|
| esv3577267 | Chr.X:135113245 - 135203184 on Build GRCh38 | Gain |
|
| dgv2433e212 | Chr.X:135163217 - 135198048 on Build GRCh38 | Gain |
|
| esv2758894 | Chr.X:135066857 - 135364390 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map