Assay Details
Target Gene Details
Entrez Gene ID: | 51032 |
Gene Name: | chymotrypsin like elastase family member 2B |
Gene Aliases: |
ELA2B |
Location: |
Chr.1:15476101-15491400 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELA2B | NM_015849.2 | NP_056933.2 | ||
| BC069412.1 | AAH69412.1 | |||
| BC069455.1 | AAH69455.1 | |||
| BC113540.1 | ||||
| BC113542.1 | ||||
| M16653.1 | AAA52381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161190 | Chr.1:15463238 - 15484851 on Build GRCh38 | Duplication |
|
| nsv441683 | Chr.1:15465929 - 15484726 on Build GRCh38 | Loss |
|
| nsv818056 | Chr.1:15470313 - 15482377 on Build GRCh38 | Loss |
|
| nsv833536 | Chr.1:15323505 - 15528830 on Build GRCh38 | Loss |
|
| esv2744319 | Chr.1:15468474 - 15488142 on Build GRCh38 | Deletion |
|
| esv3585312 | Chr.1:15468398 - 15487980 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map