Assay Details
Target Gene Details
Entrez Gene ID: | 728621 |
Gene Name: | coiled-coil domain containing 30 |
Gene Aliases: |
PFD6L, PFDN6L |
Location: |
Chr.1:42521240-42654664 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC30 | NM_001080850.2 | NP_001074319.1 | ||
| XM_011542067.2 | XP_011540369.1 | |||
| XM_011542068.2 | XP_011540370.1 | |||
| XM_011542069.2 | XP_011540371.1 | |||
| XM_011542070.2 | XP_011540372.1 | |||
| XM_011542071.2 | XP_011540373.1 | |||
| XM_017002225.1 | XP_016857714.1 | |||
| XM_017002226.1 | XP_016857715.1 | |||
| AK302075.1 | ||||
| AL832217.1 | ||||
| AY639646.1 | ||||
| BX640642.1 | CAE45789.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585850 | Chr.1:42545707 - 42568957 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142229044] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map