Assay Details
Target Gene Details
Entrez Gene ID: | 400761 |
Gene Name: | uncharacterized LOC400761 |
Gene Aliases: |
- |
Location: |
Chr.1:89624849-89632894 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLJ27354 | NR_033981.1 | |||
| AK130864.1 |
Target Gene Details
Entrez Gene ID: | 84230 |
Gene Name: | leucine rich repeat containing 8 family member C |
Gene Aliases: |
AD158, FAD158 |
Location: |
Chr.1:89615810-89719535 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC8C | XM_011542282.2 | XP_011540584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3586765 | Chr.1:89419740 - 89647558 on Build GRCh38 | Gain |
|
| esv3586769 | Chr.1:89593477 - 89694051 on Build GRCh38 | Gain |
|
| nsv830559 | Chr.1:89535847 - 89711962 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115416763] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map