Assay Details
Target Gene Details
Entrez Gene ID: | 55758 |
Gene Name: | REST corepressor 3 |
Gene Aliases: |
- |
Location: |
Chr.1:211259347-211316385 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv159013 | Chr.1:211276213 - 211280408 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map