Assay Details
Target Gene Details
Entrez Gene ID: | 6278 |
Gene Name: | S100 calcium binding protein A7 |
Gene Aliases: |
PSOR1, S100A7c |
Location: |
Chr.1:153457744-153460661 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A7 | NM_002963.3 | NP_002954.2 | ||
| AW238300.1 | ||||
| BC034687.1 | AAH34687.1 | |||
| BF090388.1 | ||||
| M86757.1 | AAA60210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831603 | Chr.1:153433366 - 153603667 on Build GRCh38 | Gain |
|
| nsv1002705 | Chr.1:153335376 - 153553113 on Build GRCh38 | Gain |
|
| nsv946399 | Chr.1:153459362 - 153465867 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map