Assay Details
Target Gene Details
Entrez Gene ID: | 163882 |
Gene Name: | consortin, connexin sorting protein |
Gene Aliases: |
C1orf71, PPP1R64 |
Location: |
Chr.1:246566337-246668587 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNST | NM_001139459.1 | NP_001132931.1 | ||
| NM_152609.2 | NP_689822.2 | |||
| XM_005273081.3 | XP_005273138.2 | |||
| XM_005273083.4 | XP_005273140.1 | |||
| XM_011544111.1 | XP_011542413.1 | |||
| XM_011544112.1 | XP_011542414.1 | |||
| XM_011544113.1 | XP_011542415.1 | |||
| XM_011544114.2 | XP_011542416.1 | |||
| XM_017000487.1 | XP_016855976.1 | |||
| XM_017000488.1 | XP_016855977.1 | |||
| XM_017000489.1 | XP_016855978.1 | |||
| AK056563.1 | BAB71218.1 | |||
| AK074142.1 | BAB84968.1 | |||
| AL834246.2 | CAD38922.2 | |||
| BC010228.2 | AAH10228.1 | |||
| BC036200.1 | AAH36200.1 | |||
| BX647629.1 | ||||
| DA461531.1 | ||||
| DA994539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv33n21 | Chr.1:246533294 - 246584284 on Build GRCh38 | Gain |
|
| esv3589483 | Chr.1:246563151 - 246571010 on Build GRCh38 | Gain |
|
| nsv1010453 | Chr.1:246482495 - 246651750 on Build GRCh38 | Gain |
|
| nsv528615 | Chr.1:246545165 - 246680013 on Build GRCh38 | Gain |
|
| esv2762191 | Chr.1:246383134 - 246651762 on Build GRCh38 | Gain |
|
| nsv428346 | Chr.1:246243844 - 246577642 on Build GRCh38 | Gain |
|
| nsv5254 | Chr.1:246550058 - 246584620 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map