Assay Details
Target Gene Details
Entrez Gene ID: | 285172 |
Gene Name: | family with sequence similarity 126 member B |
Gene Aliases: |
HYCC2 |
Location: |
Chr.2:200973718-201071671 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM126B | NM_001321618.1 | 11 | 8987 | NP_001308547.1 |
| NM_001321619.1 | 12 | 9198 | NP_001308548.1 | |
| NM_001321621.1 | 11 | 9100 | NP_001308550.1 | |
| NM_001321622.1 | 13 | 9316 | NP_001308551.1 | |
| NM_001321623.1 | 13 | 9253 | NP_001308552.1 | |
| NM_001321624.1 | 12 | 9155 | NP_001308553.1 | |
| NM_001321625.1 | 13 | 9366 | NP_001308554.1 | |
| NM_001321626.1 | 13 | 9386 | NP_001308555.1 | |
| NM_001321627.1 | 14 | 9309 | NP_001308556.1 | |
| NM_001321628.1 | 14 | 9422 | NP_001308557.1 | |
| NM_001321629.1 | 13 | 9141 | NP_001308558.1 | |
| NM_173822.4 | 12 | 9085 | NP_776183.1 |
More Information
Set Membership: |
|

Genomic Map