Assay Details
Target Gene Details
Entrez Gene ID: | 644903 |
Gene Name: | GCC2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:108507527-108534219 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCC2-AS1 | NR_135290.1 | |||
| AK095987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759081 | Chr.2:107833421 - 108606881 on Build GRCh38 | Gain |
|
| nsv834328 | Chr.2:108422875 - 108613866 on Build GRCh38 | Gain |
|
| esv2720498 | Chr.2:107831020 - 108722824 on Build GRCh38 | Deletion |
|
| nsv527491 | Chr.2:107924531 - 108650418 on Build GRCh38 | Loss |
|

Genomic Map