Assay Details
Target Gene Details
Entrez Gene ID: | 253782 |
Gene Name: | ceramide synthase 6 |
Gene Aliases: |
CERS5, LASS6 |
Location: |
Chr.2:168456249-168775134 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERS6 | NM_001256126.1 | NP_001243055.1 | ||
| NM_203463.2 | NP_982288.1 | |||
| AK172775.1 | BAD18757.1 | |||
| BC109284.1 | AAI09285.1 | |||
| BC109285.1 | AAI09286.1 | |||
| BX393696.2 |
Target Gene Details
Entrez Gene ID: | 100616356 |
Gene Name: | microRNA 4774 |
Gene Aliases: |
- |
Location: |
Chr.2:168582943-168583018 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4774 | NR_039933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428405 | Chr.2:168301712 - 168627188 on Build GRCh38 | Loss |
|
| dgv64n68 | Chr.2:168488058 - 168707865 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map