Assay Details
Target Gene Details
Entrez Gene ID: | 402117 |
Gene Name: | von Willebrand factor C domain containing protein 2-like |
Gene Aliases: |
- |
Location: |
Chr.2:214411737-214575929 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWC2L | NM_001080500.2 | NP_001073969.1 | ||
| AB374231.1 | ||||
| BC146903.1 | ||||
| BC146911.1 | ||||
| BC146931.1 | ||||
| BC146935.1 | ||||
| BC171841.1 | ||||
| EF552207.1 | ||||
| EU541473.1 |
Target Gene Details
Entrez Gene ID: | 100885780 |
Gene Name: | VWC2L intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.2:214510182-214536890 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWC2L-IT1 | NR_047698.1 | |||
| AA868132.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007833 | Chr.2:214486070 - 214546803 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78576191] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map