Assay Details
Target Gene Details
Entrez Gene ID: | 1857 |
Gene Name: | dishevelled segment polarity protein 3 |
Gene Aliases: |
DRS3 |
Location: |
Chr.3:184155311-184173614 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DVL3 | NM_004423.3 | NP_004414.3 | ||
| XM_005247172.2 | XP_005247229.1 | |||
| AF006013.1 | AAB65244.1 | |||
| AK093469.1 | ||||
| AK304686.1 | ||||
| BC032459.1 | AAH32459.1 | |||
| D86963.1 | BAA13199.2 | |||
| U49262.1 | AAB47447.1 | |||
| U75651.1 | AAB84228.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1000394 | Chr.3:184158795 - 184159977 on Build GRCh38 | Deletion |
|
| esv8912 | Chr.3:184158790 - 184159724 on Build GRCh38 | Loss |
|
| nsv1073698 | Chr.3:184159067 - 184159761 on Build GRCh38 | Deletion |
|
| nsv829812 | Chr.3:184136655 - 184302302 on Build GRCh38 | Loss |
|
| esv2514927 | Chr.3:184158393 - 184159764 on Build GRCh38 | Deletion |
|
| nsv513096 | Chr.3:184158560 - 184161350 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map