Assay Details
Target Gene Details
Entrez Gene ID: | 58477 |
Gene Name: | SRP receptor beta subunit |
Gene Aliases: |
APMCF1 |
Location: |
Chr.3:133784033-133821492 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRPRB | NM_021203.3 | NP_067026.3 | ||
| DB448771.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv513074 | Chr.3:133784364 - 133786594 on Build GRCh38 | Loss |
|
| esv3562578 | Chr.3:133784643 - 133786493 on Build GRCh38 | Deletion |
|
| esv2725943 | Chr.3:133784896 - 133786242 on Build GRCh38 | Deletion |
|
| nsv965212 | Chr.3:133784891 - 133787472 on Build GRCh38 | Duplication |
|
| nsv829729 | Chr.3:133726027 - 133901543 on Build GRCh38 | Loss |
|
| nsv4018 | Chr.3:133767023 - 133811824 on Build GRCh38 | Deletion |
|
| esv989994 | Chr.3:133784924 - 133786239 on Build GRCh38 | Deletion |
|
| esv8734 | Chr.3:133784706 - 133786378 on Build GRCh38 | Loss |
|
| esv3892 | Chr.3:133784814 - 133786372 on Build GRCh38 | Loss |
|
| esv1005045 | Chr.3:133782747 - 133792129 on Build GRCh38 | Deletion |
|
| dgv1408e212 | Chr.3:133780590 - 133789415 on Build GRCh38 | Loss |
|
| esv2661320 | Chr.3:133784922 - 133786238 on Build GRCh38 | Deletion |
|
| esv1301210 | Chr.3:133784938 - 133786254 on Build GRCh38 | Deletion |
|
| esv2833792 | Chr.3:133784927 - 133786243 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs538344066,rs569107793] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map