Assay Details
Target Gene Details
Entrez Gene ID: | 255758 |
Gene Name: | Tctex1 domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:196291219-196318294 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCTEX1D2 | NM_152773.4 | NP_689986.2 | ||
| BC021177.2 | AAH21177.1 | |||
| BM548106.1 | ||||
| CK818899.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997797 | Chr.3:195931489 - 196486087 on Build GRCh38 | Gain |
|
| nsv1009811 | Chr.3:196124925 - 196411200 on Build GRCh38 | Gain |
|
| nsv593071 | Chr.3:196132016 - 196345571 on Build GRCh38 | Gain |
|
| esv2759213 | Chr.3:195382058 - 196344722 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map