Assay Details
Target Gene Details
Entrez Gene ID: | 54729 |
Gene Name: | NK1 homeobox 1 |
Gene Aliases: |
HSPX153, NKX-1.1, SAX2 |
Location: |
Chr.4:1402932-1406442 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX1-1 | NM_001290079.1 | 2 | 852 | NP_001277008.1 |
| X76978.1 | CAA54282.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss |
|
| dgv853n67 | Chr.4:1402664 - 1408969 on Build GRCh38 | Gain |
|
| nsv4197 | Chr.4:1387665 - 1421471 on Build GRCh38 | Insertion |
|
| nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss |
|
| nsv593302 | Chr.4:1344830 - 1420896 on Build GRCh38 | Loss |
|
| nsv950333 | Chr.4:1400613 - 1419112 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map