Assay Details
Target Gene Details
Entrez Gene ID: | 644248 |
Gene Name: | SNCA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:89836401-89841991 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNCA-AS1 | NR_045481.1 | |||
| AK123890.1 | 2 | 2144 | ||
| BG181213.1 | ||||
| KF728371.1 | ||||
| KF728372.1 | ||||
| KF728373.1 | ||||
| KF728374.1 | ||||
| KF728375.1 | ||||
| KF728376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012406 | Chr.4:89246630 - 90245636 on Build GRCh38 | Loss |
|
| nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain |
|

Genomic Map