Assay Details
Target Gene Details
Entrez Gene ID: | 285456 |
Gene Name: | RPL34 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:108538190-108620457 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL34-AS1 | NR_026968.1 | |||
| AK094992.1 | BAC04469.1 | |||
| AK298931.1 | ||||
| AK308624.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3601723 | Chr.4:108565915 - 108575156 on Build GRCh38 | Gain |
|

Genomic Map