Assay Details
Target Gene Details
Entrez Gene ID: | 101929095 |
Gene Name: | uncharacterized LOC101929095 |
Gene Aliases: |
- |
Location: |
Chr.4:15004942-15427914 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929095 | NR_125911.1 | |||
| DB497821.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829868 | Chr.4:14962935 - 15163082 on Build GRCh38 | Gain |
|
| nsv461280 | Chr.4:14960872 - 15079750 on Build GRCh38 | Loss |
|

Genomic Map