Assay Details
Target Gene Details
Entrez Gene ID: | 27146 |
Gene Name: | family with sequence similarity 184 member B |
Gene Aliases: |
- |
Location: |
Chr.4:17632073-17781512 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM184B | NM_015688.1 | NP_056503.1 | ||
| AB033102.1 | ||||
| AK308144.1 |

Genomic Map