Assay Details
Target Gene Details
Entrez Gene ID: | 641700 |
Gene Name: | endothelial cell surface expressed chemotaxis and apoptosis regulator |
Gene Aliases: |
ARIA, ECSM2 |
Location: |
Chr.5:139448554-139462743 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECSCR | NM_001077693.3 | NP_001071161.1 | ||
| NM_001293739.1 | NP_001280668.1 | |||
| NR_121659.1 | ||||
| BC146912.1 | ||||
| BC146913.1 | ||||
| BI834795.1 | ||||
| DQ462572.1 | ||||
| EU025066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519808 | Chr.5:139396890 - 139805924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map