Assay Details
Target Gene Details
Entrez Gene ID: | 728586 |
Gene Name: | coiled-coil domain containing 192 |
Gene Aliases: |
LINC01183 |
Location: |
Chr.5:127702216-127941517 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC192 | NM_001317938.1 | NP_001304867.1 | ||
| XM_017009805.1 | XP_016865294.1 | |||
| XM_017009806.1 | XP_016865295.1 | |||
| XM_017009807.1 | XP_016865296.1 | |||
| XM_017009808.1 | XP_016865297.1 | |||
| XM_017009809.1 | XP_016865298.1 | |||
| XM_017009810.1 | XP_016865299.1 | |||
| XM_017009811.1 | XP_016865300.1 | |||
| XM_017009812.1 | XP_016865301.1 | |||
| XM_017009813.1 | XP_016865302.1 | |||
| HY052603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033607 | Chr.5:127049489 - 127968767 on Build GRCh38 | Gain |
|
| nsv830480 | Chr.5:127629059 - 127788830 on Build GRCh38 | Loss |
|

Genomic Map