Assay Details
Target Gene Details
Entrez Gene ID: | 54974 |
Gene Name: | tRNA-histidine guanylyltransferase 1 like |
Gene Aliases: |
ICF45, IHG-1, hTHG1 |
Location: |
Chr.5:157731389-157741449 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THG1L | NM_001317824.1 | NP_001304753.1 | ||
| NM_001317825.1 | NP_001304754.1 | |||
| NM_001317826.1 | NP_001304755.1 | |||
| NM_017872.4 | NP_060342.2 | |||
| XM_017009603.1 | XP_016865092.1 | |||
| CA426270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759389 | Chr.5:157570600 - 157806036 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map