Assay Details
Target Gene Details
Entrez Gene ID: | 10802 |
Gene Name: | SEC24 homolog A, COPII coat complex component |
Gene Aliases: |
- |
Location: |
Chr.5:134648402-134727911 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC24A | NM_001252231.1 | NP_001239160.1 | ||
| NM_021982.2 | NP_068817.1 | |||
| XM_006714523.2 | XP_006714586.1 | |||
| XM_017008961.1 | XP_016864450.1 | |||
| XM_017008963.1 | XP_016864452.1 | |||
| AJ131244.1 | CAA10334.1 | |||
| AK304060.1 | ||||
| BC019341.1 | AAH19341.1 | |||
| CD109783.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5012 | Chr.5:134646373 - 134691436 on Build GRCh38 | Deletion |
|
| nsv830491 | Chr.5:134557253 - 134724800 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map