Assay Details
Target Gene Details
Entrez Gene ID: | 153918 |
Gene Name: | zinc finger C2HC-type containing 1B |
Gene Aliases: |
C6orf94, FAM164B, dJ468K18.5 |
Location: |
Chr.6:143864436-143938346 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZC2HC1B | NM_001013623.2 | NP_001013645.1 | ||
| BC146950.1 | ||||
| BC146969.1 | ||||
| BC171862.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830830 | Chr.6:143856805 - 144059733 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map