Assay Details
Target Gene Details
Entrez Gene ID: | 6785 |
Gene Name: | ELOVL fatty acid elongase 4 |
Gene Aliases: |
ADMD, CT118, ISQMR, SCA34, STGD2, STGD3 |
Location: |
Chr.6:79914812-79947598 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL4 | NM_022726.3 | NP_073563.1 | ||
| AF277094.1 | AAG47668.1 | |||
| AK055277.1 | BAB70895.1 | |||
| AK310625.1 | ||||
| AK312511.1 | ||||
| AY037298.1 | AAK68639.1 | |||
| BC038506.1 | AAH38506.1 |
More Information
Set Membership: |
|

Genomic Map