Assay Details
Target Gene Details
Entrez Gene ID: | 2739 |
Gene Name: | glyoxalase I |
Gene Aliases: |
GLOD1, GLYI, HEL-S-74 |
Location: |
Chr.6:38675925-38703176 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLO1 | NM_006708.2 | NP_006699.2 | ||
| AB209801.1 | BAD93038.1 | |||
| AK129573.1 | ||||
| AK130312.1 | ||||
| AK293345.1 | ||||
| AK312662.1 | ||||
| AL833442.1 | ||||
| BC001741.1 | AAH01741.1 | |||
| BC011365.1 | AAH11365.1 | |||
| BC015934.1 | AAH15934.1 | |||
| BT019987.1 | AAV38790.1 | |||
| BT019988.1 | AAV38791.1 | |||
| D13315.1 | BAA02572.1 | |||
| DA342581.1 | ||||
| FJ224325.1 | ||||
| KJ534846.1 | ||||
| L07837.1 | AAA52565.1 | |||
| S83285.1 | AAB49495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830642 | Chr.6:38677280 - 38832881 on Build GRCh38 | Loss |
|
| nsv428144 | Chr.6:38621193 - 38743309 on Build GRCh38 | Gain |
|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map