Assay Details
Target Gene Details
Entrez Gene ID: | 57038 |
Gene Name: | arginyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
ArgRS, DALRD2, PCH6, PRO1992, RARSL |
Location: |
Chr.6:87513744-87590032 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RARS2 | NM_001318785.1 | NP_001305714.1 | ||
| NM_020320.4 | NP_064716.2 | |||
| NR_134857.1 | ||||
| XM_011535948.2 | 21 | 2204 | XP_011534250.1 | |
| XM_017011079.1 | XP_016866568.1 | |||
| AI610814.1 | ||||
| CA314872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6085n100 | Chr.6:87443361 - 87700332 on Build GRCh38 | Gain |
|
| nsv830722 | Chr.6:87344468 - 87513907 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map