Assay Details
Target Gene Details
Entrez Gene ID: | 10559 |
Gene Name: | solute carrier family 35 member A1 |
Gene Aliases: |
CDG2F, CMPST, CST, hCST |
Location: |
Chr.6:87472925-87512339 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35A1 | NM_001168398.1 | NP_001161870.1 | ||
| NM_006416.4 | NP_006407.1 | |||
| AJ810302.1 | CAH17780.1 | |||
| AJ810303.1 | CAH17781.1 | |||
| AJ851888.1 | CAH65468.1 | |||
| AJ851889.1 | CAH65469.1 | |||
| AJ851890.1 | CAH65470.1 | |||
| AK293696.1 | ||||
| AK299432.1 | ||||
| AL122119.1 | CAH56397.1 | |||
| BC017807.1 | AAH17807.1 | |||
| BG699364.1 | ||||
| D87969.1 | BAA13522.1 | |||
| DA210713.1 | ||||
| DB095595.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6085n100 | Chr.6:87443361 - 87700332 on Build GRCh38 | Gain |
|
| nsv830722 | Chr.6:87344468 - 87513907 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map