Assay Details
Target Gene Details
Entrez Gene ID: | 254228 |
Gene Name: | family with sequence similarity 26 member E |
Gene Aliases: |
C6orf188, dJ493F7.3 |
Location: |
Chr.6:116511645-116524792 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM26E | NM_153711.3 | 2 | 6807 | NP_714922.1 |
Target Gene Details
Entrez Gene ID: | 100128327 |
Gene Name: | trafficking protein particle complex 3 like |
Gene Aliases: |
BET3L, bA259P20.2 |
Location: |
Chr.6:116496483-116545610 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC3L | NM_001139444.2 | NP_001132916.1 | ||
| BX280366.1 | ||||
| DY655209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830783 | Chr.6:116445706 - 116645069 on Build GRCh38 | Gain |
|
| nsv523972 | Chr.6:115958024 - 116809541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map