Assay Details
Target Gene Details
Entrez Gene ID: | 493754 |
Gene Name: | RAB guanine nucleotide exchange factor 1 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.7:66530305-66592407 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GS1-124K5.11 | NR_111973.1 | |||
| AK310370.1 | ||||
| AK310548.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891135 | Chr.7:66376216 - 66777606 on Build GRCh38 | Gain |
|
| esv3613505 | Chr.7:66464247 - 66581622 on Build GRCh38 | Gain |
|

Genomic Map