Assay Details
Target Gene Details
Entrez Gene ID: | 729920 |
Gene Name: | isoprenoid synthase domain containing |
Gene Aliases: |
MDDGA7, MDDGC7, Nip, hCG_1745121 |
Location: |
Chr.7:16087527-16422562 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISPD | NM_001101417.3 | NP_001094887.1 | ||
| NM_001101426.3 | NP_001094896.1 | |||
| XM_006715770.3 | XP_006715833.1 | |||
| XM_011515500.2 | XP_011513802.1 | |||
| XM_011515508.2 | XP_011513810.1 | |||
| XM_011515509.2 | XP_011513811.1 | |||
| XM_017012577.1 | XP_016868066.1 | |||
| XM_017012578.1 | XP_016868067.1 | |||
| BE092109.1 |
More Information
Set Membership: |
|

Genomic Map