Assay Details
Target Gene Details
Entrez Gene ID: | 7586 |
Gene Name: | zinc finger with KRAB and SCAN domains 1 |
Gene Aliases: |
9130423L19Rik, KOX18, PHZ-37, ZNF139, ZNF36, ZSCAN33 |
Location: |
Chr.7:100015572-100045523 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZKSCAN1 | NM_001287054.1 | NP_001273983.1 | ||
| NM_001287055.1 | NP_001273984.1 | |||
| NM_003439.2 | NP_003430.1 | |||
| XM_005250565.4 | XP_005250622.1 | |||
| XM_011516559.2 | 1 | 393 | XP_011514861.1 | |
| AF454940.1 | AAP97699.1 | |||
| AK054676.1 | ||||
| AK074116.1 | BAB84942.1 | |||
| AK091471.1 | ||||
| AL833578.1 | ||||
| AY260738.1 | AAP83961.1 | |||
| BC022378.1 | AAH22378.1 | |||
| BX640646.1 | CAE45792.1 | |||
| HY140376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32694 | Chr.7:99967985 - 100032296 on Build GRCh38 | Loss |
|
| nsv433391 | Chr.7:99966338 - 100020983 on Build GRCh38 | Gain |
|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
| esv23518 | Chr.7:99966510 - 100027788 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map