Assay Details
Target Gene Details
Entrez Gene ID: | 6718 |
Gene Name: | aldo-keto reductase family 1, member D1 |
Gene Aliases: |
3o5bred, CBAS2, SRD5B1 |
Location: |
Chr.7:138076432-138118304 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR1D1 | NM_001190906.1 | NP_001177835.1 | ||
| NM_001190907.1 | NP_001177836.1 | |||
| NM_005989.3 | NP_005980.1 | |||
| AK058142.1 | BAB71682.1 | |||
| AK289425.1 | ||||
| AK298421.1 | ||||
| AK298428.1 | ||||
| BC130625.1 | ||||
| BC130627.1 | ||||
| FM160741.1 | ||||
| FM160742.1 | ||||
| FM160743.1 | ||||
| Z28339.1 | CAA82193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34048 | Chr.7:138039346 - 138528374 on Build GRCh38 | Loss |
|
| esv33947 | Chr.7:137235266 - 138283104 on Build GRCh38 | Gain |
|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
| nsv518070 | Chr.7:137849881 - 138124093 on Build GRCh38 | Gain |
|
| nsv831151 | Chr.7:138026701 - 138180725 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map