Assay Details
Target Gene Details
Entrez Gene ID: | 729852 |
Gene Name: | UBAP1-MVB12-associated (UMA) domain containing 1 |
Gene Aliases: |
RPA3-AS1, RPA3OS |
Location: |
Chr.7:7640636-7879223 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UMAD1 | NM_001302348.1 | NP_001289277.1 | ||
| NM_001302349.1 | NP_001289278.1 | |||
| NM_001302350.1 | NP_001289279.1 | |||
| AK295253.1 | ||||
| DA576342.1 | ||||
| DC313244.1 | ||||
| DC340488.1 | ||||
| DC354992.1 | ||||
| DC368552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6249n100 | Chr.7:6962401 - 8039172 on Build GRCh38 | Gain |
|
| esv2752177 | Chr.7:7689616 - 8138059 on Build GRCh38 | Gain |
|
| nsv8043 | Chr.7:7549564 - 7823171 on Build GRCh38 | Gain |
|
| esv3891036 | Chr.7:7491637 - 7877729 on Build GRCh38 | Gain |
|
| nsv606042 | Chr.7:7387904 - 7836835 on Build GRCh38 | Gain |
|
| esv2761144 | Chr.7:7457249 - 8757480 on Build GRCh38 | Gain+Loss |
|
| esv3891038 | Chr.7:7767046 - 7867221 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73344540] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map