Assay Details
Target Gene Details
Entrez Gene ID: | 100130771 |
Gene Name: | EF-hand calcium binding domain 10 |
Gene Aliases: |
- |
Location: |
Chr.7:105565133-105581529 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB10 | NR_027068.1 | |||
| DA101957.1 |
More Information
Set Membership: |
|

Genomic Map