Assay Details
Target Gene Details
Entrez Gene ID: | 401398 |
Gene Name: | POT1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:124929873-125145234 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POT1-AS1 | NR_125718.1 | |||
| NR_125719.1 | ||||
| AK090675.1 | BAC03501.1 | |||
| BX648695.1 | ||||
| DA063136.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6607n100 | Chr.7:124950634 - 125033319 on Build GRCh38 | Loss |
|
| nsv1016655 | Chr.7:124935630 - 125044225 on Build GRCh38 | Gain |
|
| esv2761363 | Chr.7:124951379 - 125033319 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78413916] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map