Assay Details
Target Gene Details
Entrez Gene ID: | 79783 |
Gene Name: | succinyl-CoA:glutarate-CoA transferase |
Gene Aliases: |
C7orf10, DERP13, GA3, ORF19 |
Location: |
Chr.7:40134887-40860767 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUGCT | NM_001193311.1 | NP_001180240.1 | ||
| NM_001193312.1 | NP_001180241.1 | |||
| NM_001193313.1 | NP_001180242.1 | |||
| NM_024728.2 | NP_079004.1 | |||
| XM_006715775.3 | XP_006715838.1 | |||
| XM_011515525.2 | XP_011513827.1 | |||
| XM_011515527.2 | XP_011513829.1 | |||
| XM_011515528.2 | XP_011513830.1 | |||
| XM_011515529.2 | XP_011513831.1 | |||
| XM_011515530.2 | XP_011513832.1 | |||
| XM_017012622.1 | XP_016868111.1 | |||
| AF397013.1 | AAL76418.1 | |||
| AK021870.1 | BAB13922.1 | |||
| AK296048.1 | ||||
| AK298516.1 | ||||
| AK299133.1 | ||||
| AK307935.1 | ||||
| AK308764.1 | ||||
| BC098117.1 | AAH98117.1 | |||
| BC098261.1 | AAH98261.1 | |||
| BC098310.1 | AAH98310.1 | |||
| BC098318.1 | AAH98318.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032278 | Chr.7:40057772 - 40356701 on Build GRCh38 | Gain |
|
| nsv464437 | Chr.7:40055042 - 40157569 on Build GRCh38 | Gain |
|
| esv2759525 | Chr.7:40020581 - 40229984 on Build GRCh38 | Gain |
|
| nsv971193 | Chr.7:40036282 - 40158356 on Build GRCh38 | Duplication |
|
| esv3612920 | Chr.7:40035084 - 40158562 on Build GRCh38 | Gain |
|
| nsv824080 | Chr.7:40037490 - 40159045 on Build GRCh38 | Gain |
|
| esv34547 | Chr.7:40037433 - 40191366 on Build GRCh38 | Gain |
|
| nsv1024735 | Chr.7:40109628 - 40163757 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map