Assay Details
Target Gene Details
Entrez Gene ID: | 64327 |
Gene Name: | limb development membrane protein 1 |
Gene Aliases: |
ACHP, C7orf2, DIF14, LSS, PPD2, THYP, TPT, ZRS |
Location: |
Chr.7:156673354-156893208 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMBR1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv609209 | Chr.7:156663585 - 156716626 on Build GRCh38 | Gain |
|
| nsv6020 | Chr.7:156657458 - 156687987 on Build GRCh38 | Insertion |
|
| nsv831198 | Chr.7:156629947 - 156829662 on Build GRCh38 | Loss |
|

Genomic Map