Assay Details
Target Gene Details
Entrez Gene ID: | 340351 |
Gene Name: | ATP/GTP binding protein like 3 |
Gene Aliases: |
CCP3 |
Location: |
Chr.7:134985169-135135782 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL3 | NM_178563.3 | 17 | 3116 | NP_848658.3 |
| XM_017012136.1 | 17 | 4438 | XP_016867625.1 | |
| XM_017012137.1 | 17 | 4720 | XP_016867626.1 | |
| XM_017012138.1 | 16 | 2919 | XP_016867627.1 | |
| XM_017012139.1 | 14 | 2742 | XP_016867628.1 | |
| XM_017012140.1 | 16 | 2739 | XP_016867629.1 | |
| XM_017012141.1 | 16 | 4255 | XP_016867630.1 | |
| XM_017012142.1 | 17 | 3013 | XP_016867631.1 | |
| XM_017012143.1 | 15 | 2690 | XP_016867632.1 | |
| AA707617.1 |
Target Gene Details
Entrez Gene ID: | 78996 |
Gene Name: | chromosome 7 open reading frame 49 |
Gene Aliases: |
MRI |
Location: |
Chr.7:135092303-135170826 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C7orf49 | NM_001305630.1 | NP_001292559.1 | ||
| XM_017012595.1 | XP_016868084.1 | |||
| BC010361.1 | ||||
| BC024018.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527174 | Chr.7:134952791 - 135157183 on Build GRCh38 | Loss |
|
| nsv608458 | Chr.7:135016632 - 135149118 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map