Assay Details
Target Gene Details
Entrez Gene ID: | 9988 |
Gene Name: | cyclin D binding myb like transcription factor 1 |
Gene Aliases: |
DMP1, DMTF, MRUL, hDMP1 |
Location: |
Chr.7:87152361-87196332 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMTF1 | NM_001142326.1 | NP_001135798.1 | ||
| NM_001142327.1 | NP_001135799.1 | |||
| NM_021145.3 | NP_066968.3 | |||
| NR_024549.1 | ||||
| NR_024550.1 | ||||
| XM_011516735.2 | 1 | 248 | XP_011515037.1 | |
| XM_011516737.1 | XP_011515039.1 | |||
| XM_011516739.2 | XP_011515041.1 | |||
| XM_017012865.1 | 1 | 248 | XP_016868354.1 | |
| XM_017012866.1 | 1 | 248 | XP_016868355.1 | |
| XM_017012868.1 | XP_016868357.1 | |||
| XM_017012869.1 | XP_016868358.1 | |||
| XM_017012870.1 | XP_016868359.1 | |||
| XM_017012871.1 | 1 | 248 | XP_016868360.1 | |
| XM_017012872.1 | 1 | 248 | XP_016868361.1 | |
| XM_017012873.1 | 1 | 248 | XP_016868362.1 | |
| XM_017012874.1 | XP_016868363.1 | |||
| XM_017012875.1 | 1 | 248 | XP_016868364.1 | |
| XM_017012876.1 | XP_016868365.1 | |||
| XM_017012877.1 | XP_016868366.1 | |||
| XM_017012878.1 | 1 | 248 | XP_016868367.1 | |
| AA258783.1 | ||||
| AB209230.1 | BAD92467.1 | |||
| AF084530.1 | AAC33480.1 | |||
| AF202144.1 | AAG35613.1 | |||
| AF202145.1 | AAG35614.1 | |||
| AK295144.1 | ||||
| AK296211.1 | ||||
| AK314622.1 | ||||
| BC070064.1 | AAH70064.1 | |||
| DB061156.1 | ||||
| DB069894.1 | ||||
| DC339431.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527889 | Chr.7:86866726 - 87203287 on Build GRCh38 | Gain |
|
| nsv483124 | Chr.7:87026523 - 87173407 on Build GRCh38 | Gain |
|
| nsv1016761 | Chr.7:87129228 - 87284836 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map