Assay Details
Target Gene Details
Entrez Gene ID: | 137392 |
Gene Name: | family with sequence similarity 92 member A1 |
Gene Aliases: |
- |
Location: |
Chr.8:93700252-93732041 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM92A1 | NM_001283034.1 | NP_001269963.1 | ||
| NM_145269.4 | NP_660312.2 | |||
| NR_104267.1 | ||||
| NR_104268.1 | ||||
| XM_005250787.2 | XP_005250844.1 | |||
| AK096298.1 | BAC04753.1 | |||
| BC014598.2 | AAH14598.2 | |||
| BC108707.1 | AAI08708.1 | |||
| CR627475.1 | CAH10675.1 | |||
| DQ327715.1 | ||||
| DQ327716.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831399 | Chr.8:93703175 - 93892637 on Build GRCh38 | Loss |
|
| nsv528872 | Chr.8:93677970 - 93732585 on Build GRCh38 | Loss |
|
| nsv1022473 | Chr.8:91854892 - 94250166 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map