Assay Details
Target Gene Details
Entrez Gene ID: | 138474 |
Gene Name: | TATA-box binding protein associated factor 1 like |
Gene Aliases: |
TAF(II)210, TAF2A2 |
Location: |
Chr.9:32629454-32635669 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF1L | NM_153809.2 | 1 | 4875 | NP_722516.1 |
| AF390562.1 | 1 | 4880 | AAN40840.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv614143 | Chr.9:32583509 - 33141754 on Build GRCh38 | Gain |
|
| esv3620303 | Chr.9:32569144 - 33069475 on Build GRCh38 | Gain |
|
| esv24966 | Chr.9:32628222 - 32636699 on Build GRCh38 | Gain |
|
| nsv831540 | Chr.9:32509387 - 32651746 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map