Assay Details
Target Gene Details
Entrez Gene ID: | 23392 |
Gene Name: | KIAA0368 |
Gene Aliases: |
ECM29 |
Location: |
Chr.9:111360693-111484745 on Build GRCh38 |
Assay Gene Location: | Within Intron 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0368 | NM_001080398.1 | NP_001073867.1 | ||
| XM_005251850.3 | XP_005251907.1 | |||
| XM_005251851.4 | XP_005251908.1 | |||
| XM_005251852.4 | XP_005251909.1 | |||
| XM_005251853.3 | XP_005251910.1 | |||
| XM_005251854.3 | XP_005251911.1 | |||
| XM_011518422.2 | XP_011516724.1 | |||
| XM_017014557.1 | XP_016870046.1 | |||
| AB002366.1 | BAA20823.1 | |||
| AK024705.1 | ||||
| AK025689.1 | ||||
| AK027063.1 | ||||
| AK127247.1 | ||||
| BC021127.1 | AAH21127.1 |
More Information
Set Membership: |
|

Genomic Map