Assay Details
Target Gene Details
Entrez Gene ID: | 347127 |
Gene Name: | SPATA31 subfamily D member 5, pseudogene |
Gene Aliases: |
FAM75D5, FAM75D5P |
Location: |
Chr.9:81913437-81919927 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA31D5P | NR_026851.1 | |||
| AK125938.1 | ||||
| AK128229.1 | ||||
| DB098739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831639 | Chr.9:81886633 - 82036236 on Build GRCh38 | Loss |
|
| nsv508555 | Chr.9:81892784 - 81965113 on Build GRCh38 | Deletion |
|
| nsv821665 | Chr.9:80752718 - 82292739 on Build GRCh38 | Loss |
|
| esv2738657 | Chr.9:81904006 - 81984480 on Build GRCh38 | Deletion |
|
| esv23543 | Chr.9:81912654 - 81951692 on Build GRCh38 | Gain+Loss |
|

Genomic Map