Assay Details
Target Gene Details
Entrez Gene ID: | 5524 |
Gene Name: | protein phosphatase 2 phosphatase activator |
Gene Aliases: |
PP2A, PPP2R4, PR53 |
Location: |
Chr.9:129110949-129148946 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPA | NM_001193397.1 | NP_001180326.1 | ||
| NM_001271832.1 | NP_001258761.1 | |||
| NM_021131.4 | NP_066954.2 | |||
| NM_178000.2 | NP_821067.1 | |||
| NM_178001.2 | NP_821068.1 | |||
| NM_178003.2 | NP_821070.1 | |||
| XM_011518834.2 | XP_011517136.1 | |||
| XM_011518835.2 | XP_011517137.1 | |||
| XM_011518836.2 | XP_011517138.1 | |||
| XM_011518837.2 | XP_011517139.1 | |||
| XM_011518838.2 | XP_011517140.1 | |||
| XM_017014888.1 | XP_016870377.1 | |||
| AK222788.1 | BAD96508.1 | |||
| AK297202.1 | ||||
| AK302043.1 | ||||
| AK302897.1 | ||||
| AK307424.1 | ||||
| AU125192.1 | ||||
| BC002545.2 | AAH02545.1 | |||
| BC011605.2 | AAH11605.1 | |||
| BQ896712.1 | ||||
| BT020119.1 | AAV38922.1 | |||
| CD630703.1 | ||||
| CR749810.1 | CAH18670.1 | |||
| X73478.1 | CAA51873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818725 | Chr.9:129026890 - 129176634 on Build GRCh38 | Gain |
|
| nsv831729 | Chr.9:128973042 - 129158330 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map