Assay Details
Target Gene Details
Entrez Gene ID: | 220980 |
Gene Name: | TMEM72 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:44811024-44959689 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM72-AS1 | NR_033842.1 | 8 | 2736 | |
| AK056518.1 | 8 | 2737 | ||
| DB298676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760155 | Chr.10:44707669 - 44876329 on Build GRCh38 | Gain |
|
| dgv1105n54 | Chr.10:44753495 - 44864035 on Build GRCh38 | Gain |
|
| esv3623068 | Chr.10:44713185 - 44864432 on Build GRCh38 | Gain |
|
| dgv21e55 | Chr.10:44699737 - 44861146 on Build GRCh38 | Gain |
|
| nsv831844 | Chr.10:44733343 - 44912314 on Build GRCh38 | Gain |
|
| esv32821 | Chr.10:44752266 - 44864334 on Build GRCh38 | Gain+Loss |
|
| dgv729n100 | Chr.10:44700093 - 44878497 on Build GRCh38 | Gain |
|
| nsv517161 | Chr.10:44715242 - 44920808 on Build GRCh38 | Gain+Loss |
|
| nsv831845 | Chr.10:44804820 - 44964947 on Build GRCh38 | Gain |
|
| dgv80n27 | Chr.10:44707573 - 44873696 on Build GRCh38 | Gain |
|

Genomic Map