Assay Details
Target Gene Details
Entrez Gene ID: | 253725 |
Gene Name: | family with sequence similarity 21 member C |
Gene Aliases: |
FAM21A, VPEF |
Location: |
Chr.10:45727170-45792964 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv947916 | Chr.10:45769503 - 45792769 on Build GRCh38 | Duplication |
|
| dgv114e214 | Chr.10:45766709 - 45809904 on Build GRCh38 | Gain |
|
| esv23760 | Chr.10:45766003 - 45820072 on Build GRCh38 | Gain+Loss |
|
| esv2852680 | Chr.10:45784552 - 45792552 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map