Assay Details
Target Gene Details
Entrez Gene ID: | 118932 |
Gene Name: | ankyrin repeat domain 22 |
Gene Aliases: |
- |
Location: |
Chr.10:88819899-88851975 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD22 | NM_144590.2 | NP_653191.2 | ||
| AK313969.1 | ||||
| BC021671.1 | AAH21671.1 | |||
| BM785517.1 | ||||
| DA672161.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551840 | Chr.10:88718726 - 88841108 on Build GRCh38 | Gain |
|
| nsv950991 | Chr.10:88832744 - 88854143 on Build GRCh38 | Duplication |
|
| nsv831942 | Chr.10:88804940 - 88970595 on Build GRCh38 | Gain |
|
| esv3624141 | Chr.10:88791335 - 88872446 on Build GRCh38 | Gain |
|
| esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain |
|

Genomic Map