Assay Details
Target Gene Details
Entrez Gene ID: | 22852 |
Gene Name: | ankyrin repeat domain 26 |
Gene Aliases: |
THC2, bA145E8.1 |
Location: |
Chr.10:26938195-27100498 on Build GRCh38 |
Assay Gene Location: | Within Intron 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD26 | XM_017015928.1 | XP_016871417.1 | ||
| XM_017015929.1 | XP_016871418.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2664616 | Chr.10:26927640 - 27310597 on Build GRCh38 | Deletion |
|

Genomic Map