Assay Details
Target Gene Details
Entrez Gene ID: | 7582 |
Gene Name: | zinc finger protein 33B |
Gene Aliases: |
KOX2, KOX31, ZNF11B |
Location: |
Chr.10:42574169-42638837 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF33B | NM_001305033.1 | NP_001291962.1 | ||
| NM_001305035.1 | NP_001291964.1 | |||
| NM_001305036.1 | NP_001291965.1 | |||
| NM_001305037.1 | NP_001291966.1 | |||
| NM_001305038.1 | NP_001291967.1 | |||
| NM_001305039.1 | NP_001291968.1 | |||
| NM_001305040.1 | NP_001291969.1 | |||
| NM_006955.2 | NP_008886.1 | |||
| NR_130948.1 | ||||
| NR_130949.1 | ||||
| NR_130950.1 | ||||
| NR_130951.1 | ||||
| AJ491697.1 | CAD36956.1 | |||
| AK096683.1 | ||||
| AK304306.1 | ||||
| AK309564.1 | ||||
| AK310073.1 | ||||
| BC048313.2 | AAH48313.1 | |||
| BC065181.1 | AAH65181.1 | |||
| BC107726.1 | AAI07727.1 | |||
| BG536481.1 | ||||
| CD652356.1 | ||||
| DA230792.1 | ||||
| DA493121.1 | ||||
| DA826003.1 | ||||
| DA959465.1 | ||||
| DB156590.1 | ||||
| DR000119.1 | ||||
| X68684.1 | CAA48643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3623017 | Chr.10:42584089 - 42655641 on Build GRCh38 | Loss |
|
| esv34929 | Chr.10:42620746 - 42834322 on Build GRCh38 | Loss |
|
| nsv1042764 | Chr.10:42350935 - 43016264 on Build GRCh38 | Gain |
|
| nsv550672 | Chr.10:42257742 - 42946281 on Build GRCh38 | Gain |
|
| esv2759746 | Chr.10:42469683 - 43014743 on Build GRCh38 | Loss |
|
| esv3891795 | Chr.10:42298677 - 42690427 on Build GRCh38 | Gain |
|
| nsv469725 | Chr.10:42469682 - 42682875 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115395677] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map